Order Entry
Export
ContactUsLinkComponent
 
Octyl-β-D-thioglucopyranoside
Catalog # MOLE27927684
Supplier:  Molekula
CAS Number:  
Octyl-β-D-thioglucopyranoside
Catalog # MOLE27927684
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :308.44 g/mol
Storage Temperature:Freezer
CAS Number:85618-21-9
Formula:C₁₄H₂₈O₅S
MDL Number:MFCD00012189