5,5-Dimethylacetylacetone, Sigma-Aldrich®
Catalog # SIAL697958-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:142.2 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:230-760-0
UN:1224
CAS Number:7307-04-2
Formula:C₈H₁₄O₂
MDL Number:MFCD00142767