Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dibromocarbazole
3,6-Dibromocarbazole ≥99%
3,6-Dibromocarbazole ≥99%
Irritant
3,6-Dibromocarbazole ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
3,6-Dibromocarbazole ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
6825-20-3
:
Formula:
C₁₂H₇Br₂N
MDL Number:
MFCD00004961
:
325 g/mol
Melting Pt:
204…206 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH56360
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation