Order Entry
Export
ContactUsLinkComponent
4,4'-Difluorobiphenyl
4,4'-Difluorobiphenyl
Catalog # EHERC12632015
Supplier:  EHRENSTORFER
CAS Number:  
4,4'-Difluorobiphenyl
Catalog # EHERC12632015
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :190.19 g/mol
Merck Index:13,03173
Boiling Pt:254…255 °C (1013 hPa)
Melting Pt:95 °C
Density:1.361 g/cm³ (20 °C)
EINECS:206-910-6
CAS Number:398-23-2
Formula:C₁₂H₈F₂
MDL Number:MFCD00000349