Order Entry
Export
ContactUsLinkComponent
4,4'-Dibromobiphenyl (PBB No. 15)
Irritant
4,4'-Dibromobiphenyl (PBB No. 15)
Catalog # EHERC21001500
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
4,4'-Dibromobiphenyl (PBB No. 15)
Catalog # EHERC21001500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :312 g/mol
Boiling Pt:357 °C (1013 hPa)
Melting Pt:164 °C
Density:1.867 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-198-6
UN:3152
ADR:9,II
 :GHS07
CAS Number:92-86-4
Formula:C₁₂H₈Br₂
MDL Number:MFCD00000101