1,3-Difluoro-2-nitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19064-24-5
:
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00192035
:159.09 g/mol
Boiling Pt:91…92 °C (11 mmHg)
Melting Pt:0 °C
Density:1.503
Flash Pt:88 °C (190 °F)
EINECS:242-793-8
:GHS07
Storage Temperature:Ambient