Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Naphthyridine, Sigma-Aldrich®

1,5-Naphthyridine, Sigma-Aldrich®

Glass bottle for solids, 1 g
Catalog # SIAL673218-1G
Supplier:   Merck
1,5-Naphthyridine, Sigma-Aldrich®
Catalog # SIAL673218-1G
Supplier:  Merck
CAS Number:  254-79-5
1,5-Naphthyridine, Sigma-Aldrich®

Glass bottle for solids, 1 g

CAS Number:  254-79-5
Price
129,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:254-79-5
  • Formula:C₈H₆N₂
  • MDL Number:MFCD00039727
  • Molecular Weight:130.15 g/mol
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications