ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
CAS Number:30931-67-0
:
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404
:548.68 g/mol
EINECS:250-396-6
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable: