
Alizarin red S (sodium salt), anhydrous, orange powder C.I. 58005
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:130-22-3
:93982-72-0
Formula:C₁₄H₇NaO₇S
MDL Number:MFCD00013049
:342.26 g/mol
EINECS:204-981-8
:GHS07
Storage Temperature:Ambient