Order Entry
Export
ContactUsLinkComponent
Acridine orange (chlorid salt), hydrate ∼80% (dye content), pure
Health
Acridine orange (chlorid salt), hydrate ∼80% (dye content), pure
Catalog # ACRO423340010
CAS Number:  
undefined
Health
Acridine orange (chlorid salt), hydrate ∼80% (dye content), pure
Catalog # ACRO423340010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Orange to brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 % (105°C)
  • UV spectrophotometry:
    ≥75 %

Specification Test Results

About this item

 :
Melting Pt:284…287 °C
Storage Temperature:Ambient
EINECS:200-614-0
ADR:6.1,III
 :301.81 g/mol (anhy)
 :GHS08
CAS Number:1704465-79-1
Formula:C₁₇H₂₀ClN₃·H₂O
MDL Number:MFCD00150070