Order Entry
Export
Orders LinkContactUsLinkComponent
Mitomycin C

Mitomycin C

Plastic bottle for solids, 2 mg
Catalog # ROCH10107409001
Toxic
Health
Supplier:   ROCHE
Mitomycin C
Catalog # ROCH10107409001
Supplier:  ROCHE
CAS Number:  50-07-7
Mitomycin C

Plastic bottle for solids, 2 mg

CAS Number:  50-07-7
Price
112,00 €
Log in for availability
1 * 2 mg
Product Details & Documents
Synonyms:  Mitomycin, Ametycin
  • CAS Number:50-07-7
  • Formula:C₁₅H₁₈N₄O₅
  • MDL Number:MFCD00078109
  • Molecular Weight:334.33 g/mol
  • Storage Temperature:Refrigerator
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-008-6
  • UN:2811
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2 mg

Specifications