Octyl-β-D-thioglucopyranoside
Catalog # MOLEM82602895
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:308.44 g/mol
Storage Temperature:Freezer
CAS Number:85618-21-9
Formula:C₁₄H₂₈O₅S
MDL Number:MFCD00012189