Order Entry
Export
ContactUsLinkComponent
Octyl-β-D-thioglucopyranoside ≥98%, Ultrapure
Octyl-β-D-thioglucopyranoside ≥98%, Ultrapure
Catalog # J575-1G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Octyl-β-D-thioglucopyranoside ≥98%, Ultrapure
Catalog # J575-1G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Purity:
    ≤98%
  • Solubility (5%, water):
    Pass
  • Specific Rotation (1%, MeOH):
    (- 55) – (-49°)

Specification Test Results

About this item

 :
 :308.44 g/mol
Storage Temperature:Freezer
CAS Number:85618-21-9
Formula:C₁₄H₂₈O₅S
MDL Number:MFCD00012189