Order Entry
Export
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) (from fish sperm)
DNA (Deoxyribonucleic acid) (from fish sperm)
Catalog # ROCH11467140001
Supplier:  ROCHE
CAS Number:  
DNA (Deoxyribonucleic acid) (from fish sperm)
Catalog # ROCH11467140001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:309-566-6
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922