Order Entry
Export
ContactUsLinkComponent
n-Undecyl-β-D-maltoside
n-Undecyl-β-D-maltoside
Catalog # MOLE16709684-1G
Supplier:  Molekula
CAS Number:  
n-Undecyl-β-D-maltoside
Catalog # MOLE16709684-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:170552-39-3
Formula:C₂₃H₄₄O₁₁