Order Entry
Export
ContactUsLinkComponent
n-Octyl-β-D-maltoside, Sigma-Aldrich®
n-Octyl-β-D-maltoside, Sigma-Aldrich®
Catalog # SIAL19181-1G
Supplier:  Merck
CAS Number:  
n-Octyl-β-D-maltoside, Sigma-Aldrich®
Catalog # SIAL19181-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
CAS Number:82494-08-4
Formula:C₂₀H₃₈O₁₁