Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Octyl-β-D-maltoside
n-Octyl-β-D-maltoside
n-Octyl-β-D-maltoside
n-Octyl-β-D-maltoside
Catalog # MOLE90033684
Supplier:
Molekula
CAS Number:
Expand image
n-Octyl-β-D-maltoside
Catalog # MOLE90033684
Supplier:
Molekula
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Specifications
About this item
Product Details
:
CAS Number:
82494-08-4
Formula:
C₂₀H₃₈O₁₁
Product Details