Order Entry
Export
Orders LinkContactUsLinkComponent
n-Dodecyl-β-D-maltoside

n-Dodecyl-β-D-maltoside

Irritant
n-Dodecyl-β-D-maltoside
Certificates
n-Dodecyl-β-D-maltoside

3 Options Available Below

Product Details & Documents
Synonyms:  DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
  • CAS Number:69227-93-6
  • Formula:C₂₄H₄₆O₁₁
  • MDL Number:MFCD00043012
  • Molecular Weight:510.6 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g