n-Decyl-β-D-maltoside
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
About this item
Synonyms: DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside
- CAS Number:82494-09-5
- Formula:C₂₂H₄₂O₁₁
- MDL Number:MFCD00061624
- Molecular Weight:482.56 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside
- CAS Number:82494-09-5
- Formula:C₂₂H₄₂O₁₁
- MDL Number:MFCD00061624
- Molecular Weight:482.56 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



