Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
ROCH10109886001
:
ROCHE
:
Soy bean trypsin inhibitor
ROCH10109886001
:
ROCHE
:
Pack type:
Plastic bottle for solids
Pk:
50 mg
CAS No.:
9035-81-8
Species:
Soy bean
Storage conditions:
Freezer
Protein/peptide name:
Trypsin inhibitor