Trypsin inhibitor
Plastic bottle for solids, 1 g
Catalog # ROCH10109878001
Supplier: ROCHE
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 g
- Cat. No.:ROCH10109878001
- Protein/peptide name:Trypsin inhibitor
- Storage conditions:Freezer
- CAS No.:9035-81-8



