Pepstatin A
Plastic bottle for solids, 50 mg
Catalog # ROCH11524488001
Supplier: ROCHE
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 mg
- Cat. No.:ROCH11524488001
- Molecular formula:C₃₄H₆₃N₅O₉
- Protein/peptide name:Pepstatin A
- Storage conditions:Refrigerator
- MDL No.:MFCD00060740
- CAS No.:26305-03-3



