Order Entry
Export
Orders LinkContactUsLinkComponent
Pepstatin A

Pepstatin A

Plastic bottle for solids, 2 mg
Catalog # ROCH10253286001
Supplier:   ROCHE
Pepstatin A
Catalog # ROCH10253286001
Supplier:  ROCHE
CAS Number:  
Pepstatin A

Plastic bottle for solids, 2 mg

CAS Number:  
Price
113,00 €
Log in for availability
1 * 2 mg
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2 mg
  • Cat. No.:
    ROCH10253286001
  • Molecular formula:
    C₃₄H₆₃N₅O₉
  • Protein/peptide name:
    Pepstatin A
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00060740
  • CAS No.:
    26305-03-3

Specifications