Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Pepstatin A
Pepstatin A
Pepstatin A
Supplier:
BIOMOL RESEARCH LABORATORIES
Certificates
Expand image
Pepstatin A
Supplier:
BIOMOL RESEARCH LABORATORIES
About this item
Specifications
CAS No.:
26305-03-3
Storage conditions:
Refrigerator
MDL No.:
MFCD00060740
Protein/peptide name:
Pepstatin A
Molecular formula:
C₃₄H₆₃N₅O₉
Specifications