Leupeptin hemisulphate
Plastic bottle for solids, 25 mg
Catalog # ROCH11017128001
Supplier: ROCHE
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 mg
- Cat. No.:ROCH11017128001
- Molecular formula:C₄₀H₇₈N₁₂O₁₂S
- Protein/peptide name:Leupeptin hemisulphate
- Storage conditions:Freezer
- Protein synonyms:(Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
- MDL No.:MFCD00037012
- CAS No.:103476-89-7



