Actinomycin D (from Streptomyces species) ≥95% (by HPLC) cell culture reagent, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-76-0
:
Formula:C₆₂H₈₆N₁₂O₁₆
MDL Number:MFCD00005033
:1255.5 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ300
UN:3462