Order Entry
Export
ContactUsLinkComponent
 
7-Aminoactinomycin D (7-AAD)
Catalog # 40037.
Supplier:  Biotium
CAS Number:  
7-Aminoactinomycin D (7-AAD)
Catalog # 40037.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1270.43 g/mol
CAS Number:7240-37-1
Formula:C₆₂H₈₇N₁₃O₁₆
MDL Number:MFCD00077295