Cyclosporin A
FITZ10C66M6032524
:
- Pack type:Glass bottle for solids
- Pk:1 mg
- Source:
- Species:
- Environmentally Preferable:
- Storage conditions:Refrigerator
- Protein/peptide name:Cyclosporin A
Signal word: Danger
GHS Symbols: GHS08, GHS07
Signal word: Danger
GHS Symbols: GHS08, GHS07