Order Entry
Export
ContactUsLinkComponent
Nystatin
Irritant
Nystatin
Catalog # PROOICRS0369
CAS Number:  
Irritant
Nystatin
Catalog # PROOICRS0369
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg
  • Seq:
    2530

Specifications

About this item

 :
 :926.09 g/mol
EINECS:215-749-0
 :GHS07
Storage Temperature:Freezer
CAS Number:1400-61-9
Formula:C₄₇H₇₅NO₁₇
MDL Number:MFCD25541651