Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amphotericin B
Amphotericin B, 850 U/mg
Amphotericin B, 850 U/mg
Irritant
Amphotericin B, 850 U/mg
Supplier:
Roth Carl
Certificates
Expand image
Irritant
Amphotericin B, 850 U/mg
Supplier:
Roth Carl
About this item
CAS Number:
1397-89-3
:
Formula:
C₄₇H₇₃NO₁₇
MDL Number:
MFCD00877763
:
924.08 g/mol
EINECS:
215-742-2
:
GHS07
Storage Temperature:
Freezer