
Amphotericin B (from Streptomyces species), yellow powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1397-89-3
:
Formula:C₄₇H₇₃NO₁₇
MDL Number:MFCD00877763
:924.08 g/mol
Melting Pt:>170 °C (lit.)
EINECS:215-742-2
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Potency:≥800 µg/mg
- Source:Streptomyces sp.
- Expiration Date:Month/Day/Year