Order Entry
Export
ContactUsLinkComponent
Amphotericin B
Irritant
Amphotericin B
Catalog # EHERC10243050
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Amphotericin B
Catalog # EHERC10243050
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :924.08 g/mol
EINECS:215-742-2
 :GHS07
Storage Temperature:Freezer
CAS Number:1397-89-3
Formula:C₄₇H₇₃NO₁₇
MDL Number:MFCD00877763