Malt extract
Glass bottle for solids, 500 g
Catalog # ROTHX976.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:8002-48-0
- MDL Number:MFCD00131570
- Storage Temperature:Ambient
- EINECS:232-310-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



