Order Entry
Export
ContactUsLinkComponent
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate 95%
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate 95%
Catalog # ACRO226225000
CAS Number:  
undefined
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate 95%
Catalog # ACRO226225000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 % (105°C, 5 h)
  • HPLC:
    ≥94 %

Specification Test Results

About this item

 :
 :527.12 g/mol (anhy)
Melting Pt:215…218 °C
Storage Temperature:Freezer
EINECS:252-849-3
CAS Number:652154-13-7
 :81012-87-5
Formula:C₉H₁₄N₃Na₂O₁₄P₃·xH₂O
MDL Number:MFCD00150809