

Thymidine (2'-deoxythymidine), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-89-5
:146183-25-7
Formula:C₁₀H₁₄N₂O₅
MDL Number:MFCD00006537
:242.23 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-070-4
:GHS08
:GHS07
Storage Temperature:Ambient