Order Entry
Export
Orders LinkContactUsLinkComponent
 

Adenosine, Sigma-Aldrich®

Supplier:   Merck
Adenosine, Sigma-Aldrich®
Supplier:  Merck
Certificates
Adenosine, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Refrigerator
  • EINECS:200-389-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 36,80 €
    36,80 €
    1 * 5 g
       36,80 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 125,00 €
    125,00 €
    1 * 25 g
       125,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 388,00 €
    388,00 €
    1 * 100 g
       388,00 €