Adenosine
Glass bottle for solids, 1 g
Catalog # ROTH2318.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
- CAS Number:58-61-7
- Formula:C₁₀H₁₃N₅O₄
- MDL Number:MFCD00005752
- Molecular Weight:267.24 g/mol
- Storage Temperature:Refrigerator
- EINECS:200-389-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



