Adenosine 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
- CAS Number:58-61-7
- Formula:C₁₀H₁₃N₅O₄
- MDL Number:MFCD00005752
- Molecular Weight:267.24 g/mol
- Melting Pt:233…238 °C
- Storage Temperature:Ambient
- EINECS:200-389-9
4 Options Available Below
Product Details & Documents
Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
- CAS Number:58-61-7
- Formula:C₁₀H₁₃N₅O₄
- MDL Number:MFCD00005752
- Molecular Weight:267.24 g/mol
- Melting Pt:233…238 °C
- Storage Temperature:Ambient
- EINECS:200-389-9
Documents
ACRO16404
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:233 to 238 °C
- Loss on drying:≤0.5 % (105 °C, 2 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≥99.0 %
- UV spectrophotometry:98.0 to 102.0 % (lambda max. 259 nm, pH 7)
- UV transmittance:≥98 % (c=0.5, H2O, 430 nm) 1 cm cell
- Ratio of Absorbances:0.76 to 0.80 (A250/A260)
- Nitrogen (N):25.69 to 26.73 % (on dry basis)
- Heavy metals (as Pb):≤10 ppm



