Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloroadenosine
2-Chloroadenosine, Sigma-Aldrich®
2-Chloroadenosine, Sigma-Aldrich®
2-Chloroadenosine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2-Chloroadenosine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
146-77-0
:
Formula:
C₁₀H₁₂ClN₅O₄
MDL Number:
MFCD00005734
:
301.69 g/mol
EINECS:
205-678-3