Chromotrope 2R
Glass bottle for solids, 50 g
Catalog # ROTH4937.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:4197-07-3
- Formula:C₁₆H₁₀N₂Na₂O₈S₂
- MDL Number:MFCD00003955
- Molecular Weight:468.37 g/mol
- EINECS:224-085-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



