MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid)
Catalog # MOLE59874779-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Hydroxy-3-morpholinopropane-1-sulphonic acid
:
:225.27 g/mol
EINECS:269-989-6
:GHS07
Storage Temperature:Ambient
CAS Number:68399-77-9
Formula:C₇H₁₅NO₅S
MDL Number:MFCD00009629