1,2-Bis(diphenylphosphino)benzene, Sigma-Aldrich®
Catalog # SIAL460273-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:446.46 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:13991-08-7
Formula:C₃₀H₂₄P₂
MDL Number:MFCD00014081