Order Entry
Export
ContactUsLinkComponent
Creatine phosphate disodium salt ≥97%, high purity
Creatine phosphate disodium salt ≥97%, high purity
Catalog # 0271-25G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Creatine phosphate disodium salt ≥97%, high purity
Catalog # 0271-25G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • ATP:
    ≤0.002%
  • Inorganic Phosphate:
    ≤0.5%
  • Purity (Enzymatic):
    >97%
  • Water (KF):
    20 - 24%

Specification Test Results

About this item

 :
 :255.077 g/mol
Storage Temperature:Freezer
EINECS:213-074-6
CAS Number:922-32-7
 :19333-65-4
Formula:C₄H₈N₃O₅P·2Na
MDL Number:MFCD00014351