Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
Supplier:
G-Biosciences
Certificates
Expand image
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
Supplier:
G-Biosciences
About this item
CAS Number:
85261-19-4
:
Formula:
C₁₆H₃₃NO₆
Storage Temperature:
Ambient