Order Entry
Export
Orders LinkContactUsLinkComponent
Brilliant green, certified

Brilliant green, certified

Bottle, amber, 100 g
  0502-100G
Irritant
Brilliant green, certified
  0502-100G
 :  633-03-4
Brilliant green, certified
Brilliant green, certified

Bottle, amber, 100 g

 :  633-03-4
 
272,00 €
Log in for availability
1 * 100 g
 

A stain for differentation of E.coli from S.typhosa. Also used in bacteriological media. Colour index: 42040

Synonyms:  Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
  • CAS Number:633-03-4
  • Formula:C₂₇H₃₄N₂O₄S
  • MDL Number:MFCD00011880
  • Molecular Weight:482.65 g/mol
  • Storage Temperature:Ambient
  • EINECS:211-190-1
Specifications

Specification Test Results

  • Absorbance @ lambda Max:
    ≥0.550
  • Dye Content:
    ≥85.0%
  • Lambda max (.003 mg/ml 50% Ethanol):
    628 – 632 nm

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    100 g

Specifications