Brilliant green
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 633-03-4 Molecular Formula: C27H34N2O4S Formula Weight: 482.65 Melting Point: 210° dec. Beilstein Reference: 3901207 Merck Reference: 14,1374 MDL No.: MFCD00011880
Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
- CAS Number:633-03-4
- Formula:C₂₇H₃₄N₂O₄S
- MDL Number:MFCD00011880
- Molecular Weight:482.65 g/mol
- Merck Index:14,1374
- Melting Pt:210 °C (dec.)
- Storage Temperature:Ambient
- EINECS:211-190-1
2 Options Available Below
Product Details & Documents
CAS No.: 633-03-4 Molecular Formula: C27H34N2O4S Formula Weight: 482.65 Melting Point: 210° dec. Beilstein Reference: 3901207 Merck Reference: 14,1374 MDL No.: MFCD00011880
Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
- CAS Number:633-03-4
- Formula:C₂₇H₃₄N₂O₄S
- MDL Number:MFCD00011880
- Molecular Weight:482.65 g/mol
- Merck Index:14,1374
- Melting Pt:210 °C (dec.)
- Storage Temperature:Ambient
- EINECS:211-190-1



