Organic reference standard, 3,3'-Dichlorobenzidine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL12377900AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:91-94-1
- Description:3,3'-Dichlorobenzidine 10 µg/ml in acetonitrile
- Formula:C₁₂H₁₀Cl₂N₂
- Description:3,3'-Dichlorobenzidine