2,2'-Bipyridine-4,4'-dicarboxylic acid
Catalog # MOLEM85271529
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.20 g/mol
CAS Number:6813-38-3
Formula:C₁₂H₈N₂O₄
MDL Number:MFCD00015430