Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Bipyridine-4,4'-dicarboxylic acid
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%
Irritant
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
6813-38-3
:
Formula:
C₁₂H₈N₂O₄
MDL Number:
MFCD00015430
:
244.20 g/mol
Melting Pt:
>310 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB22594
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation