
2,2'-Bipyridine-4,4'-dicarboxylic acid 98%
Catalog # ACRO363660010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Titration excess NaOH:≥97.5 %
Specification Test Results
About this item
:
:244.20 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:6813-38-3
Formula:C₁₂H₈N₂O₄
MDL Number:MFCD00015430